Calcium acetate xhydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:114460-21-8
:5743-26-0
Formula:(H₃CCOO)₂Ca·nH₂O
MDL Number:MFCD00150019
:176.18 g/mol
Merck Index:14,1646
Melting Pt: Dec.
Density:1,5
Storage Temperature:Ambient
EINECS:200-540-9