Cell culture medium, Neuropan basal medium
Catalog # PANBP04-00904
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Neuropan basal medium, without L-arginine and L-lysine
Specifications
About this item
Neuropan basal medium supports the growth of hippocampus cells and many other neuronal cells of the central nervous system.
- Stored at 2 to 8 °C
A feeder layer of astrocytes is not required. Neuropan basal medium does not contain glutamate which should be added for the initial culture (25 μM). Before use, Neuropan basal medium is supplemented with serum or for a serum-free culture with Neuropan 27 or NS21 Supplement.