Order Entry
Singapore
ContactUsLinkComponent
Glucose solution
Glucose solution
Catalog # PANBP04-01701
Supplier:  PAN BIOTECH
CAS Number:  
Glucose solution
Catalog # PANBP04-01701
Supplier:  PAN BIOTECH
CAS Number:  
Specifications
  • Pk:
    100 mL
  • Description:
    Glucose solution, 100 g/L, sterile
Specifications
About this item
Glucose solution essential for cell culture media.
  • Autoclaved
Stored at room temperature.