Foetal bovine serum, FBS Premium, South American origin
PANBP30-1901
- Sterility:Sterile filtered
- Description:Foetal bovine serum, FBS Premium, South America origin, heat inactivated
- Pk:100 mL
This serum is heated for 30 min to 56 °C in a water bath under repeated gentle mixing.
- 0,2 µm sterile filtered
- Store at ‒20 °C
Application includes:
Measurements of lactate dehydrogenase in the culture supernatant as a marker for cell damage (serum LDH is inactivated by heat)
Minimises lot-to-lot variations in serum (all thermolabile components are removed)
Studies on vitamins and growth factors
Enhance viral safety, since heat-labile viruses are inactivated
Tests that do not tolerate presence of complement (complement destruction)