Cell culture media, Endopan 300 SL
PANBP04-0065K
:
- Media Format:Liquid
- Pk:500 mL
- Description:Endopan 300 SL kit, with 9 supplements
- Shelf life:3 Months
Endopan 300 SL kit is provided with serum substitute and supplements in separate sterile packing. This will enable the user to prepare a medium for special applications.
- Serum-free medium for HUVEC and endothelial cells
- Store at 2 to 8 °C
: This kit contains the following components in single packing:
Basal medium Endopan 300 SL
Serum substitute Panexin SL-S
EGF (human recombinant epidermal growth factor)
FGF-2 (human recombinant basic fibroblast growth factor)
VEGF (human recombinant vascular endothelial growth factor)
Vitamin C (ascorbic acid phosphate)
R3-IGF-1 (human recombinant insulin-like growth factor)
GA (gentamycin/amphotericin)
Hydrocortisone
Heparin
Basal medium Endopan 300 SL
Serum substitute Panexin SL-S
EGF (human recombinant epidermal growth factor)
FGF-2 (human recombinant basic fibroblast growth factor)
VEGF (human recombinant vascular endothelial growth factor)
Vitamin C (ascorbic acid phosphate)
R3-IGF-1 (human recombinant insulin-like growth factor)
GA (gentamycin/amphotericin)
Hydrocortisone
Heparin
: For research use only. Not approved for human or animal diagnostic or therapeutic procedures.