Order Entry
Singapore
Orders LinkContactUsLinkComponent
Freezing media
Marketsource
Freezing media
Marketsource
Freezing media
Freezing medium, cryopreservation-medium for the cryoconservation of cells. The medium is based on DMEM, supplemented with a mix of fetal bovine serum and DMSO.
  • Stored at −20 °C
This composition guarantees a high survival rate and excellent cell growth after thawing.
Product Family Options
  • Description
    Pk
  • Freezing medium, cryopreservation-medium, with DMSO, with FBS
    10 mL
    1 * 10 mL
    1 * 10 mL
    Marketsource
    Marketsource
  • Freezing medium, cryopreservation-medium, with DMSO, with FBS
    50 mL
    1 * 50 mL
    1 * 50 mL - $473.00
    $473.00
    1 * 50 mL
       $473.00
    Marketsource
    Marketsource
  • Freezing medium, with 8,7% DMSO with 0,1% methyl cellulose
    50 mL
    1 * 50 mL
    1 * 50 mL
    Marketsource
    Marketsource