Order Entry
Singapore
ContactUsLinkComponent
Hydrocortisone
  PANBP07-15010
 :  
Hydrocortisone
  PANBP07-15010
 :  
  • Pk:
    10 mL
  • Description:
    Hydrocortisone, 1 mg/ml in Ethanol
Hydrocortisone tested according to the highest possible quality standards.
  • Stored at −20 °C
Dissolved according to in-house specifications, sterilised and filtered at 0,2 μm.