Demecolcine solution 10 µg/ml
Catalog # PANBP07-91050
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:371.433 g/mol
Storage Temperature:Freezer
CAS Number:477-30-5
Formula:C₂₁H₂₅NO₅
MDL Number:MFCD00075459