Cell separation solutions, Pancoll
Supplier: PAN BIOTECH
Certificates
About this item
The isolation of cells is the first step for gene expression studies or in diagnostic procedures. Besides biological separation techniques physical separation methods are most commonly used. These methods use physical differences such as size and weight of the particles to be separated. For this purpose so-called separating solutions (= centrifugation media) are used.
- When properly stored, separating solutions are stable for at least 36 months
- The storage period starts with the manufacturing date
- Steriled
- Store at +2 °C to room temperature
Pancoll separating solutions contain a polysaccharide with a molecular weight of 400000 daltons; this hydrophilic polymer allows for production of aqueous solutions for cell separation with a density of up to 1,2 g/ml. PAN-Biotech offers a variety of ready-to-use products with a density of 1,063 g/ml up to 1,091 g/ml for a very wide range of cell separation applications.