Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl sulphoxide
Dimethyl sulphoxide cell culture reagent endotoxin tested
Dimethyl sulphoxide cell culture reagent endotoxin tested
Dimethyl sulphoxide cell culture reagent endotoxin tested
PANBP60-36750100
:
PAN BIOTECH
:
Dimethyl sulphoxide cell culture reagent endotoxin tested
PANBP60-36750100
:
PAN BIOTECH
:
Pk:
100 mL
Seq:
2440
:
:
78.14 g/mol
:
Ambient
:
67-68-5
:
(CH₃)₂SO
:
MFCD00002089