Order Entry
Singapore
ContactUsLinkComponent
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%
Catalog # PANBP08-33001
Supplier:  PAN BIOTECH
CAS Number:  
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%
Catalog # PANBP08-33001
Supplier:  PAN BIOTECH
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :414.33 g/mol
CAS Number:298-93-1
Formula:C₁₈H₁₆N₅BrS
MDL Number:MFCD00011964