Serum-free media, Panserin 293S
Supplier: PAN BIOTECH
Certificates
About this item
Panserin 293S is a complete ready to use medium for the serum-free cultivation of HEK293 cells (Human Embryonic Kidney) in suspension culture.
- Store at 2 to 8 °C
Based on DMEM/F12 medium additional trace elements, cholesterol and herbal hydrolysates have been added. It does not contain any proteins or
components of animal or human origin.
It is a particularly enriched medium optimised for the growth of HEK293 cells in suspension culture and quickly provides high cell densities.