Cell culture media, BSK-H
PANBP04-25300S1
:
- Media Format:Liquid
- Pk:500 mL
- Description:BSK-H medium without L-glutamine, with 2,2 g/L sodiumbicarbonate, with 6% rabbit serum
BSK-H medium is a standardised complex medium designed to support the growth of the Lyme disease spirochete, Borrelia burgdorferi.