Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Thymidine (2'-Deoxythymidine)
Thymidine (2'-deoxythymidine) 100X (1.6 mM)
Thymidine (2'-deoxythymidine) 100X (1.6 mM)
Thymidine (2'-deoxythymidine) 100X (1.6 mM)
PANBP04-56020
:
PAN BIOTECH
:
Thymidine (2'-deoxythymidine) 100X (1.6 mM)
PANBP04-56020
:
PAN BIOTECH
:
Pk:
20 mL
Seq:
2305
:
:
242.23 g/mol
:
Refrigerator
:
50-89-5
:
146183-25-7
:
C₁₀H₁₄N₂O₅
:
MFCD00006537