Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Kanamycin sulphate
Kanamycin sulphate 10 mg/ml
Kanamycin sulphate 10 mg/ml
Health
Kanamycin sulphate 10 mg/ml
Supplier:
PAN BIOTECH
Certificates
Expand image
Health
Kanamycin sulphate 10 mg/ml
Supplier:
PAN BIOTECH
About this item
CAS Number:
25389-94-0
:
Formula:
C₁₈H₃₈N₄O₁₅S
MDL Number:
MFCD00070253
:
582.6 g/mol
:
GHS08
Storage Temperature:
Freezer