Cell culture medium, Optipan, basal medium Eagle (BME)
PANBP04-08501
:
- Media Format:Liquid
- Pk:500 mL
- Description:Optipan, basal medium MEM eagle with EBBS, with L-glutamine, with 2,2 g/L sodium carbonate
- Shelf life:12 Months
Optipan is a ready to use complete medium for the serum free or serum reduced cultivation of a variety of cells.
- Sterile
- Store from 2 to 8 °C
- Contains no undefined peptones or hydrolysates
The addition of attachment factors allows the cultivation of even highly demanding cells after a short adaptaion phase.