Order Entry
Singapore
Orders LinkContactUsLinkComponent
Tris buffer, pH 8,4

Tris buffer, pH 8,4

Tris buffer, 1 M, 8,4, 500 mL
Catalog # PANBP05-05500
Marketsource
Supplier:   PAN BIOTECH
Tris buffer, pH 8,4
Catalog # PANBP05-05500
Supplier:  PAN BIOTECH
Marketsource
CAS Number:  
Tris buffer, pH 8,4

Tris buffer, 1 M, 8,4, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
  • Sterile
  • Store at 2 to 8 °C
Specifications

Specifications

  • Concentration:
    1 M
  • Pk:
    500 mL
  • Description:
    Tris buffer
  • pH value:
    8,4

Specifications