Cell dissociation solution, Accumax
Supplier: PAN BIOTECH
Certificates
About this item
Accumax can be used as a direct replacement of trypsin for cell detachment or collagenase for cell dissociation or as a anti-clumping agent for cell culture, flow cytometry, and FACS.
- Ready-to-use sterile solution
- Neutralising of the enzymes is not required for routine cell culture passaging
- Active at room temperature, no pre-warming required or recommended
- Preserves most epitopes for subsequent flow cytometry analysis
Gentle detachment of cells for analysis of cell surface markers, transfection procedures, migration or proliferation assays, flow cytometry, and routine cell passage.
Stored at −20 °C (in dark), thawed at 2 to 8 °C (up to 2 months).
Specifications
- Shelf life:2 years
- Composition:Accumax enzymes (activity 1200 - 2000 U/ml) in DPBS, without Mg/Ca, with 0,5 mM EDTA