Starch 0,5% in aqueous solution stabilised, TECHNICAL
VWRC4729.0500
:
- Pack type:Plastic bottle
- Pk:500 mL
- Seq:2725
- Environmentally Preferable:
- Identification:Passes test
:
:Stabilizer
:162.14 g/mol
:Ambient
:232-679-6
:9005-25-8
:(C₆H₁₀O₅)n
:MFCD00082026