Dulbecco's phosphate buffered saline (DPBS)
The main purpose of buffered salt solutions is the maintenance of pH and osmolarity in cell culture applications as well as providing essential inorganic ions.
- Stored at 2 °C to room temperature
These solutions are usually used for washing tissues and cells. Often they are combined with other agents in cell and tissue treatment.