Order Entry
Singapore
ContactUsLinkComponent
Cell culture supplements, Panexin CD, serum replacement
Cell culture supplements, Panexin CD, serum replacement
Catalog # PANBP04-93100
Supplier:  PAN BIOTECH
CAS Number:  
Cell culture supplements, Panexin CD, serum replacement
Catalog # PANBP04-93100
Supplier:  PAN BIOTECH
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Panexin CD, serum replacement with defined components

Specifications

About this item

Panexin CD is a chemically defined serum replacement for the cultivation of adherent and non-adherent cells under serum-free culture conditions or to significantly reduce the amount of serum in cell culture. It supports the growth of many cell types in an optimum manner without any extra handling compared to serum.

  • Sterile solution
  • Store at ‒20 °C, in the dark
  • Stability: two years from date of production

Panexin CD contains recombinant proteins, lipids, salts, amino acids, trace elements, hormones and a 3-dimensional substrate release system in an optimised formulation. It contains no growth factors, undefined hydrolysates or peptones.

Panexin CD is designed to replace or to reduce serum in the cell culture in a very simple manner. Compared to other serum replacements, it contains only chemically defined components. Due to its very defined character, lot testing is not necessary anymore. It also allows high reproducibility and simplified downstream processing.

Panexin CD enables defined proliferation and differentiation of stem cells. Characterisation studies of growth factors will obtain more reproducible and clearer results. Panexin CD is also useful to develop sensitive cell-based in vitro tests and coculture procedures. For cell lines which require specific growth factors these should be added in a concentration as previously used.

Caution: For research use only. Not approved for human or animal diagnostic or therapeutic procedures.