Cell culture media, Glasgow MEM BHK 21
Glasgow MEM (GMEM) was originally formulated for growing BHK 21 cells and it was developed as a modification of BME to culture primary baby hamster kidney cells.
- Sterile media
This version has twice the concentration of vitamins and amino acids.
- Storage temperature:2...8 °C