L(+)-Glutamic acid, powder Ph. Eur., USP, pharma grade
Catalog # PANBP60-17041000
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:147.13 g/mol
Storage Temperature:Ambient
CAS Number:56-86-0
Formula:HO₂CCH₂CH₂CH(NH₂)CO₂H
MDL Number:MFCD00002634