Order Entry
Singapore
ContactUsLinkComponent
EDTA ferric monosodium salt
EDTA ferric monosodium salt
Catalog # VWRC280233V
Supplier:  VWR Chemicals
CAS Number:  
undefined
EDTA ferric monosodium salt
Catalog # VWRC280233V
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Assay (EDTA):
    66 - 71 %
  • Assay (calculated as Fe(III)):
    12.5 - 14.0 %
  • Insolubility in water:
    Max. 0.1 %
  • Cl (Chloride):
    Max. 0.05 %
  • SO4 (Sulphate):
    Max. 0.2 %
  • As (Arsenic):
    Max. 3 ppm
  • Pb (Lead):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :367.05 g/mol
Merck Index:12,04076
Melting Pt:80 °C
EINECS:239-802-2
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:15708-41-5
Formula:C₁₀H₁₂FeN₂NaO₈
MDL Number:MFCD00078215