Order Entry
Singapore
ContactUsLinkComponent
Sodium fluoride 87% ≥87% (w/v) in sodium hydroxide solution
Irritant
Sodium fluoride 87% ≥87% (w/v) in sodium hydroxide solution
Catalog # VWRC19817DVEMPTY
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Sodium fluoride 87% ≥87% (w/v) in sodium hydroxide solution
Catalog # VWRC19817DVEMPTY
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    100 L
  • Seq:
    5429

Specifications

Specification Test Results

  • Appearance:
    Clear Colourless Liquid
  • Sodium hydroxide (W/V):
    0.0198 - 0.0242%
  • Fluoride (W/V):
    1.300 - 1.365%

Specification Test Results

About this item

 :
 :41.99 g/mol
EINECS:231-667-8
 :GHS07
CAS Number:7681-49-4
Formula:NaF
MDL Number:MFCD00003524