Filtrated bovine serum, FBS good, Australia origin
Supplier: PAN BIOTECH
Certificates
About this item
FBS Good is a naturally defined serum with a sustained growth promoting property and a higher safety.
- Reproducible growth properties
- Very low endotoxin level
- Suitable for a great variety of cells
- Continuous quality control
- 0,2 µm sterile filtered
- Store at ‒20 °C
FBS Good only contains serum of highest quality from defined countries as specified. It is not blended or enhanced by addition of growth factors or proteins.