Bovine serum albumin (BSA)
Catalog # PANBP06-139405
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:5 g
- Source:Serum
- Species:Bovine
- Storage conditions:Powder and preserved solutions at 2...8 °C; preservative free solution at –20 °C
- Form:Lyophilised powder
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Purity:>99%
Specifications
About this item
Albumins serve as additive proteins for tissue cultures. They are the main protein component in serum and are added to cell culture media to increase the stability of cell membranes and to bind possibly toxic elements.
- Minimal lot to lot variation
- Stringent control of raw materials
- Minimal BSE/TSE risk
- US origin
- Full documentation
- Special products for unique applications