About this item
ENDOPAN MV ready-to-use is a specially developed medium for the in vitro culture of human microvascular endothelial cells (MV-EC) containing all components necessary for optimal growth. It is designed for use in an incubator at 37 °C with a 5% CO₂ atmosphere.
- Sterile medium
- Store basal or complete medium at 2 to 8 °C, store supplements at ‒20 °C
ENDOPAN MV Kit is provided with FBS and supplements in separate sterile packing. This will enable the user to prepare a medium for special applications. For example, FBS, VEGF, FGF-2, or other components may be omitted from the complete medium for specific experimental settings.
Note: Such a formulation will not promote optimal cell growth. Therefore, this composition can not be used for routine long-term culture of endothelial cells. Please make sure that sterility is not compromised when adding individual components to prepare complete medium. The medium should be carefully but thoroughly mixed after addition of all components to assure a homogeneous solution.