Serum-free media, Panserin 412
Catalog # PANBP04-710412
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Panserin 412, with L-Glutamine
Specifications
About this item
Panserin 412 is a fully defined, complete, ready-to-use medium for the serum-free cultivation of a multitude of adherent cells.
- Multi-purpose medium suitable for a variety of adherent cells
- Store at 2 to 8 °C
Based on Iscove's MEM, trace elements, albumin, cholesterol, soya lipids and vitamins and were added to the medium. It does not contain any growth factors.
It contains special attachment factors for the successful cultivation of cells that hardly attach. With every adaption to serum-free media, changes of the cells should be taken into consideration. These changes may concern the morphology, the karyotype, the surface marker etc. Thus cells in serum-free medium don't always have to be identical with those from the culture containing serum in which they originate (selection).
Caution: For research purpose only. Not suitable for human or animal diagnostic or therapeutics procedures.