Order Entry
Singapore
ContactUsLinkComponent
Serum-free media, Panserin 411
Serum-free media, Panserin 411
Catalog # PANBP04-710411
Supplier:  PAN BIOTECH
CAS Number:  
Serum-free media, Panserin 411
Catalog # PANBP04-710411
Supplier:  PAN BIOTECH
CAS Number:  

Specifications

  • Media Format:
    Liquid
  • Pk:
    500 mL
  • Description:
    Panserin 411, with L-Glutamine

Specifications

About this item

Panserin 411 is a fully defined, complete ready-to-use medium for the serum-free cultivation of a multitude of adherent and non adherent cells which are Insulin-dependent (e.g. CHO-cells).

  • Multi-purpose medium suitable for a variety of cells
  • Store at 2 to 8 °C

In adherent as well as non adherent cells can be cultivated. As the medium contains no growth factors there is a possibility to investigate the effects of specific growth factors added to the cell culture. It does not contain any attachment factors. With some cell types a pre-treatment of the cell culture vessels with gelatine, collagen, poly-D-lysine or fibronectin may support or enable a culture under serum-free conditions.

With every adaption to serum-free media, changes of the cells should be taken into consideration. These changes may concern morphology, karyotype, surface markers and so on. Thus cells in serum-free medium may not be identical with those from cultures containing serum in which they originated (selection).

Caution: For research purpose only. Not suitable for human or animal diagnostic or therapeutics procedures.