VWR®, Fuel testing standards - jet aviation fuel
Supplier: VWR Chemicals
Certificates
About this item
Fuel testing Certified Reference Materials are for the verification of laboratory instruments used in testing petroleum and derivative products. This includes, but is not limited to, the analysis and measurement of: Acidity content, sulphur content, aniline point, Fluorescent Indicator Adsorption (FIA) aromatics, freezing point.
- Highest level of accreditation guaranteed, providing the most credible data currently available worldwide
- Low levels of uncertainty achieves maximum accuracy of data
- All data generated exclusively by ISO 17025 accredited independent laboratories
- Ensures good inter-laboratory correlation
- Certified in accordance with ASTM / EN or IP test method protocols
- Fully traceable to international standards
- Tamper-evident security packaging
Certifications: Fully certified in accordance with ISO 17034 under accreditation by UKAS.
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberSDS_EN_SG_VWRC99122.pdf SDS_EN_SG_VWRC99124.pdf SDS_EN_SG_VWRC99127.pdf sds_VWRC99122 sds_VWRC99124
SDS
More
VWR® for Calibrations