Order Entry
Singapore
ContactUsLinkComponent
Digital timer, four channels, with white board/pen, Lab Alert®
Digital timer, four channels, with white board/pen, Lab Alert®
Catalog # HEATHS120365
Supplier:  HEATHROW
undefined
Digital timer, four channels, with white board/pen, Lab Alert®
Catalog # HEATHS120365
Supplier:  HEATHROW

Specifications

  • Colour:
    White
  • Description:
    Digital timer, 4 channels, with white board and pen, 24 hour
  • W×D×H:
    123×86×24 mm
  • Pk:
    1

Specifications

About this item

Clock features count up or down to 23 hours 59 minutes. It stands up or attaches to a magnetic surface (magnets on back of unit).

  • Four timer channels
  • Sounds when time expires
  • 10 mm display digits
  • Up/down count direction
Delivery: Whiteboard, marking pen and batteries (two AAA) included.