VWR®, 4-Place Mini Bead Mill Homogeniser
Catalog # VWRI432-0366
Supplier: VWR Collection
Specifications
- Capacity:Max. 4 tubes: 4×0,5 ml / 4×1,5 ml / 4×2 ml / 4×7 ml
- Description:4-place Mini Bead Mill homogeniser, 230 V
- Seq:001
- Environmentally Preferable:
- Model:4-place Mini Bead Mill
- Application areas:Sample preparation, DNA, RNA, protein extraction
- Electrical:100 - 230 V, 50/60 Hz
- Speed range:1 - 5 m/s
- Sample volume:25 µl - 5 ml
- Run time:1 s - 5 min
- Weight:7,48 kg
- W×D×H:215×292×254 mm
- Pk:1
Specifications
About this item
The VWR® 4-place Mini Bead Mill homogeniser is compact and powerful.
- Specifically designed for grinding, lysing and the homogenisation of biological samples prior to molecular extraction
- Ideal solution for releasing DNA, RNA proteins and enzymes from tough samples, while still retaining molecular integrity
- Offers a broad performance range
Using sample tubes pre-filled with a variety of lysing beads, the mill vigorously and uniformly shakes the tubes providing an efficient, consistent high yield and quality homogenisation in less than 60 seconds. The VWR® 4-place Mini Bead Mill homogeniser supports simultaneous processing of samples in 4×0,5, 4×1,5, 4×2 or 1×7 ml tubes. It does not require a cool down period between runs, allowing non stop use and high throughput. The unit features a convenient front loading design and safety lid closure.
Delivery: Drive unit is supplied with integrated 0,5, 1,5, and 2 ml tube carriage, interchangeable 1×7 ml tube carriage, user manual and 2 year warranty. VWR® disposable bead tubes must be ordered separately.