Adjustable volume pipettes, Pearl®
: HEATHROW
The Pearl® is a single-channel or multi-channel pipette offering adjustable volume and precision liquid handling.
- Internal metal parts for increased accuracy and calibration stability, and long life
- Easy volume adjustment with locking tab to prevent accidental change
- Separate tip ejector
- Large digit volume display
- Colour-coded and labelled with volume range
- Both top and side have colour-coded components for quick visual confirmation
- Autoclavable
The sleek, ergonomic design, reduced plunger pressure and slight downward slant to tip ejector help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires two-third the operation pressure of most standard pipettes.
: Each pipette is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.