Order Entry
Singapore
ContactUsLinkComponent
 
 
 
 
Methanol ≥99.8% for HPLC, Rankem®
 :  RANKEM
 
undefined
 
 
 
Methanol ≥99.8% for HPLC, Rankem®
 :  RANKEM
 

 

  • Color (APHA):
    ≤ 10
  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.80 %
  • Weight Per ml (at 20°C):
    0.790 -- 0.792 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.0005 %
  • Titrable Acid:
    Max. 0.0002 meq/g
  • Titrable Base:
    Max. 0.0002 meq/g
  • Water ( By KF):
    Max. 0.05 %
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 220 nm:
    Min. 55 %
  • 240 nm:
    Min. 85 %
  • From 260 nm:
    Min. 98 %