Aquapoint Karl Fischer Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
Karl Fischer titration is a widely used analytical method for
water determination, which works over a wide range from
1 ppm to 100% and provides reproducible and accurate
results. Rankem Aquapoint is used for water determination in
pharmaceuticals, chemicals, food & beverages sample.
- Improved safety
- Free from pyridine, chlorinated solvents, ethanolamine and methanol
- Accurate & Reproducible results
- Long storage stability
- Convenient, Universal application
- Applicable for analysis of wide range of solids & liquid (including aldehyde & Ketone) samples using suitable buffer/working medium
- Sharp & stable end point (5 - 6 mg /ml)
Brown coloured solution free from suspension
Specifications
- UN No.:2924
- Flash point:39 °C (closed cup)