



n-Heptane ≥99.00%, Laboratory Reagent, Rankem®
RANKH2045
: RANKEM
:
- Pack type:Amber coloured glass bottle
- Pk:2,5 L
- Seq:5096
- Guarantee Analysis:.
- Assay(GC):Min. 99.00 %
- Weight Per ml (at 20°C):0.683 --> 0.684 g
- Color:≤ 10 APHA
- Maximum Limits Of Impurities:.
- Residue On Evaporation:Max. 0.005 %
- Water ( By KF):Max. 0.05 %
:
:100.20 g/mol
:98,4 °C (1013 hPa)
:–90,6 °C
:0,6838 g/cm³ (20 °C)
:−4 °C
:Ambient
:205-563-8
:1206
:3,II
:GHS02
:GHS08
:GHS07
:GHS09
:142-82-5
:H₃C(CH₂)₅CH₃
:MFCD00009544