Leucolyse RUBY for hematology, J.T.Baker®
Catalog # BAKR2989.5000PC
Supplier: Avantor
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Conductivity (uS):100 - 180
- Osmolality (mOsm/kg):65 - 75
- Performance Test:passes test
- pH at 25°C:8.3 - 8.7
- at 265-275 nm:0.400 - 0.600
Specification Test Results
About this item
:
Storage Temperature:Refrigerator
:1