Order Entry
Singapore
ContactUsLinkComponent
 
Dimethyl sulphoxide ≥99.90% for gas chromatography, Rankem®
Catalog # RANKD0176
Supplier:  RANKEM
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.90% for gas chromatography, Rankem®
Catalog # RANKD0176
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    Amber coloured glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Color (APHA):
    ≤ 10
  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.90 %
  • Weight Per ml (at 20°C):
    1.099 --> 1.101 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.0005 %
  • Titrable Acid:
    Max. 0.0002 meq/g
  • Water ( By KF):
    Max. 0.05 %

Specification Test Results

About this item

 :
 :78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18,5 °C
Density:1,101 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
REACH:01-2119431362-50
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089