Order Entry
Singapore
ContactUsLinkComponent
Dimethyl sulphoxide ≥99.8% for HPLC, Rankem®
Dimethyl sulphoxide ≥99.8% for HPLC, Rankem®
Catalog # RANKD0171
Supplier:  RANKEM
CAS Number:  
undefined
Dimethyl sulphoxide ≥99.8% for HPLC, Rankem®
Catalog # RANKD0171
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pack type:
    Amber colored glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Colour (APHA):
    ≤10
  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.80%
  • Weight Per ml (at 20 °C):
    1.099 --> 1.101 g
  • Refractive Index (at 20 °C):
    1.478 --> 1.479
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.001%
  • Titrable Acid:
    Max. 0.0002 meq/g
  • Titrable Base:
    Max. 0.0002 meq/g
  • Water ( By KF):
    Max. 0.10%
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 270 nm:
    Min. 35%
  • 280 nm:
    Min. 60%
  • 300 nm:
    Min. 80%
  • 330 nm:
    Min. 90%
  • From 350 nm:
    Min. 98%

Specification Test Results

About this item

 :
 :78.14 g/mol
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089