


N,N-Dimethylformamide ≥99.90% for gas chromatography, Rankem®
Catalog # RANKD0134
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Amber colored glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Colour (APHA):≤10
- Guarantee Analysis:.
- Assay(GC):Min. 99.90%
- Weight Per ml (at 20 °C):0.947 --> 0.949 g
- Identity (IR):Conforms
- Maximum Limits Of Impurities:.
- Residue After Evaporation:Max. 0.0005%
- Water (By KF):Max. 0.05%
Specification Test Results
About this item
:
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–60,5 °C
Density:0,949 g/cm³ (20 °C)
Flash Pt:57,5 °C (DIN 51755) (closed cup)
EINECS:200-679-5
UN:2265
ADR:3,III
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:68-12-2
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284