



Cyclohexane ≥99.7% for HPLC, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:110-82-7
:
Formula:C₆H₁₂
MDL Number:MFCD00003814
:84.16 g/mol
Melting Pt:7 °C
Flash Pt:–18 °C
UN:1145
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient