Order Entry
Singapore
ContactUsLinkComponent
Methanol ≥99.9% gradient gold for HPLC, Rankem®
Methanol ≥99.9% gradient gold for HPLC, Rankem®
Catalog # RANKM0351
Supplier:  RANKEM
CAS Number:  
undefined
Methanol ≥99.9% gradient gold for HPLC, Rankem®
Catalog # RANKM0351
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

About this item

 :
 :32.04 g/mol
Melting Pt:–98 °C
Density:0,7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
UN:1230
 :GHS02
 :GHS06
 :GHS08
Storage Temperature:Ambient
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595