Order Entry
Singapore
ContactUsLinkComponent
 
Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™
  HONC679178-4X4L
 :  
Water with 0,05% (v/v) formic acid, CHROMASOLV™ LabReady™, Solvent Blend for HPLC, Riedel-de Haën™
  HONC679178-4X4L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

 

 

  • Contains:
    0.05 %(w/v) ammonium formate

 

 

 :
 :100 °C
 :0,9986 g/cm³ (20 °C)
 :Ambient
 :231-791-2
 :7732-18-5
 :64-18-6
 :MFCD00011332