

2-Propanol ≥99.00% USP, Macron Fine Chemicals™
MACR3593-08
:
- Pk:200 L
- Seq:5549
- USP - Description:Passes Test
- USP - Solubility:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25 °C:0.783 - 0.787
- USP - Refractive Index at 20 °C:1.3760 - 1.3780
- USP - Acidity:Passes Test
- USP - Diethyl Ether:Max. 0.1%
- USP - Acetone:Max. 0.1%
- USP - Diisopropyl Ether:Max. 0.1%
- USP - n-Propyl alcohol:Max. 0.1%
- USP - 2-butanol:Max. 0.1%
- USP - Total:Max. 1.0%
- USP - Nonvolatile Residue:≤0.005%
- USP - Assay:≥99.00%
- USP- Water:≤0.5%
:
:60.1 g/mol
:12 °C
:Ambient
:1219
:3,II
:GHS02
:GHS07
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674