

Acetic acid glacial ≥99.8% for HPLC, Rankem®
Catalog # RANKA0042
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Amber coloured glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- BIS Specification:.
- Guarantee Analysis:.
- Relative Density at 27/ 27°C:≤ 1.0483
- Color (Hazen):≤ 20
- Acetic Acid (CH3COOH):≥ 99.70 %
- Crystallizing point:≥ 16 °C
- Maximum Limits Of Impurities:.
- Residue On Evaporation:≤ 0.001 %
- Chloride (Cl):<= 1 ppm
- Iron (Fe):<= 1 ppm
- Sulfate (SO4):<= 1 ppm
- HEAVY METALS [(including iron) calculated as Pb]:<= 2 ppm
- Formic acid (HCOOH):≤ 0.025 %
- Acetaldehyde (CH3CHO):≤ 0.003 %
- Oxidizable impurities:Passes test
- Water content:≤ 0.20 %
- HPLC Specification:.
- Assay:≥ 99.80 %
- Identity (IR):Conforms
- Weight Per ml (at 20°C):1.048 - 1.050 g
- UV Transmittance:.
- Wavelength:.
- 255 nm:≥ 30 %
- 260 nm:≥ 80 %
- from 290 nm:≥ 98 %
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:17 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:38,5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS02
:GHS05
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152