


Acetone ≥99.8% for HPLC, Rankem®
: RANKEM
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:56,2 °C (1013 hPa)
:–95,4 °C
:0,792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:GHS02
:GHS07
:GHS08
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Purity:≥ 99.70%
- Relative Density at 27/27°C:0.784 - 0.786
- Color:≤ 10 APHA
- Residue On Evaporation, mg/100 ml:≤ 5.00
- Water content:≤ 0.40%
- Acidity (As CH₃COOH) g/100 ml:≤ 0.002
- Alkalinity:Passes test
- Permanganate Test:≥ 30 min
- Alcoholic Impurities:Passes test
- Benzene content:≤ 2.00 mg/kg
- Methanol content:≤ 300.0 mg/kg
- Assay (GC):≥ 99.80 %
- Weight Per ml (at 20 °C):0.790 - 0.791 g
- Identity (IR):Conforms
- Residue On Evaporation:≤ 0.0003%
- Titrable Acid:≤ 0.0003 meq/g
- Titrable Base:≤ 0.0003 meq/g
- Water (By KF):≤ 0.10 %
- Wavelength:UV Transmittance
- 330 nm:≥ 15 %
- 335 nm:≥ 50 %
- 340 nm:≥ 80 %
- From 350 nm:≥ 98 %