

Acetonitrile ≥99.8% for preparative HPLC, Rankem®
Catalog # RANKA2104
Supplier: RANKEM
CAS Number:
Specifications
- Pack type:Amber colored glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Guarantee Analysis:.
- Assay(GC):≥ 99.80 %
- Weight Per ml (at 20°C):0.782 - 0.783 g
- Maximum Limits Of Impurities:.
- Acidity (As CH3COOH):≤ 0.0015 %
- Water ( By KF):≤ 0.05 %
- Non volatile matter:≤ 0.005 %
- UV Transmittance:.
- 220 nm:≥ 85 %
- 230 nm:≥ 92 %
- 240 nm:≥ 98 %
Specification Test Results
About this item
:
:41.05 g/mol
Merck Index:13,00071
Boiling Pt:81,6 °C (1013 hPa)
Melting Pt:–45,7 °C
Density:0,782 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-835-2
UN:1648
ADR:3,II
:Filtered through a 0.2 µm filter.
REACH:01-2119471307-38
:GHS02
:GHS07
CAS Number:75-05-8
Formula:H₃CCN
MDL Number:MFCD00001878