Sodium acetate, anhydrous ≥99% FCC, USP, J.T.Baker®
Catalog # BAKR3473-X1
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Bag
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Kit Components Meet Requirements:Passes Test
Specification Test Results
About this item
:
:82.03 g/mol
Merck Index:12,08711
Melting Pt:324 °C
:Management of Change (MOC) category = R
Storage Temperature:Ambient
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00012459