Order Entry
Singapore
ContactUsLinkComponent
Toluene ≥99.8% for HPLC, Rankem®
Flammable
Health
Irritant
Toluene ≥99.8% for HPLC, Rankem®
Supplier:  RANKEM
Certificates
undefined
Flammable
Health
Irritant
Toluene ≥99.8% for HPLC, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Guarantee Analysis:
    .
  • Assay:
    Min. 99.80 %
  • Weight Per ml (at 20°C):
    0.865 --> 0.867 g
  • Maximum Limits Of Impurities:
    .
  • Non volatile matter:
    Max. 0.0005 %
  • Acidity/Alkalinity:
    Max. 0.0004 %
  • Substance Discoloured By H2SO4:
    Passes test
  • Water ( By KF):
    Max. 0.03 %
  • UV Transmittance:
    .
  • Wavelength:
    .
  • 285 nm:
    Min. 10 %
  • 310 nm:
    Min. 79 %
  • 320 nm:
    Min. 90 %
  • 350 nm:
    Min. 98 %

Specification Test Results

Related Information